2,4-Dichloropyridine
Bottle, 100 g
Catalog # APOSOR6843-100G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
- CAS Number:26452-80-2
- Formula:C₅H₃Cl₂N
- MDL Number:MFCD00955618
- Molecular Weight:147.99 g/mol
- Boiling Pt:189…190 °C
- Melting Pt:–1 °C
- Density:1.37 g/cm³ (25 °C)
- Flash Pt:89 °C
- Storage Temperature:Ambient
- EINECS:247-717-7
Specifications
Specifications
- Pack type:Bottle
- Pk:100 g



