3,5-Dimethoxybenzaldehyde 97%
Bottle, 500 g
Catalog # APOSOR4981-500G
Supplier: Apollo Scientific
3,5-Dimethoxybenzaldehyde 97%
Catalog # APOSOR4981-500G
Supplier: Apollo Scientific
CAS Number: 7311-34-4
Jump to:
Product Details & Documents
- CAS Number:7311-34-4
- Formula:C₉H₁₀O₃
- MDL Number:MFCD00003366
- Molecular Weight:166.18 g/mol
- Boiling Pt:115…118 °C/1mmHg
- Melting Pt:44…48 °C
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:500 g



