1,4-Benzodioxane
Bottle, 100 g
APOSOR4961-100G
: 2,3-Dihydrobenzo[b][1,4]dioxine, 2,3-Dihydro-1,4-benzodioxin, Benzo-1,4-dioxane, 1,4-Benzodioxan
- :493-09-4
- :C₈H₈O₂
- :MFCD00006821
- :136.15 g/mol
- :216 °C
- :1.17 g/cm³ (25 °C)
- :87 °C
- :Ambient
- :207-775-6
- Pack type:Bottle
- Pk:100 g