Order Entry
Export
Orders LinkContactUsLinkComponent
1,9-Dibromononane

1,9-Dibromononane

Bottle, 25 g
Catalog # APOSOR5434-25G
Irritant
Supplier:   Apollo Scientific
1,9-Dibromononane
Catalog # APOSOR5434-25G
Supplier:  Apollo Scientific
CAS Number:  4549-33-1
1,9-Dibromononane

Bottle, 25 g

CAS Number:  4549-33-1
Price
24,60 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Nonamethylene dibromide
  • CAS Number:4549-33-1
  • Formula:BrH₂C(CH₂)₇CH₂Br
  • MDL Number:MFCD00000279
  • Molecular Weight:286.05 g/mol
  • Boiling Pt:285…288 °C
  • Melting Pt:–2 °C
  • Density:1.407 g/cm³ (25 °C)
  • Flash Pt:113 °C
  • Storage Temperature:Ambient
  • EINECS:224-913-0
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications