3,4-Dichlorophenylacetylene
Bottle, 1 g
Catalog # APOSOR5185-1G
Supplier: Apollo Scientific
3,4-Dichlorophenylacetylene
Catalog # APOSOR5185-1G
Supplier: Apollo Scientific
CAS Number: 556112-20-0
Jump to:
Product Details & Documents
Synonyms: 1,2-Dichloro-4-ethynylbenzene
- CAS Number:556112-20-0
- Formula:C₈H₄Cl₂
- MDL Number:MFCD07783046
- Molecular Weight:171.023 g/mol
- Melting Pt:40…44 °C
- Flash Pt:96.1 °C
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:1 g
- Cat. No.:APOSOR5185-1G



