1,1-Dimethylurea
Bottle, 100 g
Catalog # APOSOR48247-100G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: N,N-Dimethylurea
- CAS Number:598-94-7
- Formula:C₃H₈N₂O
- MDL Number:MFCD00007959
- Molecular Weight:88.11 g/mol
- Melting Pt:178…183 °C
- Density:1.255 g/cm³ (25 °C)
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:100 g



