Order Entry
Export
Orders LinkContactUsLinkComponent
 

D-(+)-Xylose

Bottle, 100 g
Catalog # APOSOR7967-100G
Irritant
Supplier:   Apollo Scientific
D-(+)-Xylose
Catalog # APOSOR7967-100G
Supplier:  Apollo Scientific
CAS Number:  58-86-6
D-(+)-Xylose

Bottle, 100 g

CAS Number:  58-86-6
Price
21,80 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:58-86-6
  • Formula:C₅H₁₀O₅
  • MDL Number:MFCD00151475
  • Molecular Weight:150.13 g/mol
  • Melting Pt:147…158 °C
  • Density:1.525 g/cm³ (25 °C)
  • Flash Pt:>110 °C
  • Storage Temperature:Ambient
  • EINECS:200-400-7

 

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications