Order Entry
Export
Orders LinkContactUsLinkComponent
1,4-Dimethylpiperazine

1,4-Dimethylpiperazine

Bottle, 1 kg
Catalog # APOSOR5144-1KG
Flammable
Corrosive
Supplier:   Apollo Scientific
1,4-Dimethylpiperazine
Catalog # APOSOR5144-1KG
Supplier:  Apollo Scientific
CAS Number:  106-58-1
1,4-Dimethylpiperazine

Bottle, 1 kg

CAS Number:  106-58-1
Price
246,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  sym-Dimethylpiperazine
  • CAS Number:106-58-1
  • Formula:C₆H₁₄N₂
  • MDL Number:MFCD00006156
  • Molecular Weight:114.19 g/mol
  • Boiling Pt:131…132 °C/750mmHg
  • Melting Pt:–1 °C
  • Density:0.844 g/cm³ (25 °C)
  • Flash Pt:18 °C
  • Storage Temperature:Ambient
  • EINECS:203-412-0
  • UN:2924
  • ADR:3+8,II
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 kg

Specifications