Order Entry
Export
Orders LinkContactUsLinkComponent
Imidazole

Imidazole

Bottle, 2,5 kg
Catalog # APOSOR5102-2.5KG
Corrosive
Irritant
Health
Supplier:   Apollo Scientific
Imidazole
Catalog # APOSOR5102-2.5KG
Supplier:  Apollo Scientific
CAS Number:  288-32-4
Imidazole

Bottle, 2,5 kg

CAS Number:  288-32-4
Price
78,20 €
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  Glyoxaline, 1H-Imidazole, Glyoxalamine
  • CAS Number:288-32-4
  • Formula:C₃H₄N₂
  • MDL Number:MFCD00005183
  • Molecular Weight:68.08 g/mol
  • Boiling Pt:235 °C
  • Melting Pt:85…91 °C
  • Flash Pt:145 °C
  • Storage Temperature:Ambient
  • EINECS:206-019-2
  • UN:3263
  • ADR:8,II

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    2,5 kg

Specifications