Imidazole
Bottle, 1 kg
Catalog # APOSOR5102-1KG
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: Glyoxaline, 1H-Imidazole, Glyoxalamine
- CAS Number:288-32-4
- Formula:C₃H₄N₂
- MDL Number:MFCD00005183
- Molecular Weight:68.08 g/mol
- Boiling Pt:235 °C
- Melting Pt:85…91 °C
- Flash Pt:145 °C
- Storage Temperature:Ambient
- EINECS:206-019-2
- UN:3263
- ADR:8,II
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Bottle
- Pk:1 kg



