Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Diphenylacetone

1,3-Diphenylacetone

Bottle, 100 g
Catalog # APOSOR52322-100G
Supplier:   Apollo Scientific
1,3-Diphenylacetone
Catalog # APOSOR52322-100G
Supplier:  Apollo Scientific
CAS Number:  102-04-5
1,3-Diphenylacetone

Bottle, 100 g

CAS Number:  102-04-5
Price
43,50 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  1,3-Diphenylpropan-2-one, 1,3-Diphenyl-2-propanone
  • CAS Number:102-04-5
  • Formula:(C₆H₅)CH₂C(O)CH₂(C₆H₅)
  • MDL Number:MFCD00004795
  • Molecular Weight:210.28 g/mol
  • Boiling Pt:330 °C
  • Melting Pt:32…34 °C
  • Density:1.04 g/cm³ (25 °C)
  • Flash Pt:113 °C
  • Storage Temperature:Ambient
  • EINECS:203-000-0
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications