Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,5-Dimethylindole

Bottle, 25 g
Catalog # APOSOR5331-25G
Corrosive
Irritant
Supplier:   Apollo Scientific
2,5-Dimethylindole
Catalog # APOSOR5331-25G
Supplier:  Apollo Scientific
CAS Number:  1196-79-8
2,5-Dimethylindole

Bottle, 25 g

CAS Number:  1196-79-8
Price
78,20 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  2,5-Dimethyl-1H-indole
  • CAS Number:1196-79-8
  • Formula:C₁₀H₁₁N
  • MDL Number:MFCD00005621
  • Molecular Weight:145.2 g/mol
  • Melting Pt:113…114 °C
  • Storage Temperature:Ambient

 

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications