Order Entry
Export
Orders LinkContactUsLinkComponent
2-Chloro-4,5-difluoroaniline

2-Chloro-4,5-difluoroaniline

Bottle, 25 g
Catalog # APOSPC302442-25G
Irritant
Health
Supplier:   Apollo Scientific
2-Chloro-4,5-difluoroaniline
Catalog # APOSPC302442-25G
Supplier:  Apollo Scientific
CAS Number:  2613-32-3
2-Chloro-4,5-difluoroaniline

Bottle, 25 g

CAS Number:  2613-32-3
Price
52,10 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:2613-32-3
  • Formula:C₆H₄ClF₂N
  • MDL Number:MFCD08448325
  • Molecular Weight:163.55 g/mol
  • Melting Pt:55…58 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications