Order Entry
Export
Orders LinkContactUsLinkComponent
4,7-Dihydroxyquinazoline

4,7-Dihydroxyquinazoline

Bottle, 1 g
Catalog # APOSOR471457-1G
Supplier:   Apollo Scientific
4,7-Dihydroxyquinazoline
Catalog # APOSOR471457-1G
Supplier:  Apollo Scientific
CAS Number:  16064-25-8
4,7-Dihydroxyquinazoline

Bottle, 1 g

CAS Number:  16064-25-8
Price
108,00 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:16064-25-8
  • Formula:C₈H₆N₂O₂
  • MDL Number:MFCD01823180
  • Molecular Weight:162.145 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications