Order Entry
Export
Orders LinkContactUsLinkComponent
 

7-Nitroindole

Bottle, 5 g
Catalog # APOSOR6214-5G
Irritant
Health
Supplier:   Apollo Scientific
7-Nitroindole
Catalog # APOSOR6214-5G
Supplier:  Apollo Scientific
CAS Number:  6960-42-5
7-Nitroindole

Bottle, 5 g

CAS Number:  6960-42-5
Price
23,20 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:6960-42-5
  • Formula:C₈H₆N₂O₂
  • MDL Number:MFCD00005683
  • Molecular Weight:162.15 g/mol
  • Melting Pt:96…97 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications