Order Entry
Export
Orders LinkContactUsLinkComponent
7-Nitroindole

7-Nitroindole

Bottle, 25 g
Catalog # APOSOR6214-25G
Irritant
Health
Supplier:   Apollo Scientific
7-Nitroindole
Catalog # APOSOR6214-25G
Supplier:  Apollo Scientific
CAS Number:  6960-42-5
7-Nitroindole

Bottle, 25 g

CAS Number:  6960-42-5
Price
81,10 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:6960-42-5
  • Formula:C₈H₆N₂O₂
  • MDL Number:MFCD00005683
  • Molecular Weight:162.15 g/mol
  • Melting Pt:96…97 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications