Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,6-Pyridinedimethanol

Bottle, 100 g
Catalog # APOSOR6456-100G
Corrosive
Irritant
Supplier:   Apollo Scientific
2,6-Pyridinedimethanol
Catalog # APOSOR6456-100G
Supplier:  Apollo Scientific
CAS Number:  1195-59-1
2,6-Pyridinedimethanol

Bottle, 100 g

CAS Number:  1195-59-1
Price
264,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  2,6-Bis(hydroxymethyl)pyridine
  • CAS Number:1195-59-1
  • Formula:C₇H₉NO₂
  • MDL Number:MFCD00006351
  • Molecular Weight:139.15 g/mol
  • Boiling Pt:185 °C/15mmHg
  • Melting Pt:110…115 °C
  • Storage Temperature:Ambient
  • EINECS:214-803-0
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications