Lithium bromide 99%
Bottle, 100 g
APOSIN2314-100G
Jump to:
- CAS Number:7550-35-8
- Formula:LiBr
- MDL Number:MFCD00011077
- Molecular Weight:86.84 g/mol
- Boiling Pt:1265 °C
- Melting Pt:550 °C
- Density:3.46 g/cm³ (25 °C)
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:100 g



