Order Entry
Export
Orders LinkContactUsLinkComponent
Iron(III) bromide, anhydrous

Iron(III) bromide, anhydrous

Bottle, 5 g
Catalog # APOSIN2056-5G
Irritant
Supplier:   Apollo Scientific
Iron(III) bromide, anhydrous
Catalog # APOSIN2056-5G
Supplier:  Apollo Scientific
CAS Number:  10031-26-2
Iron(III) bromide, anhydrous

Bottle, 5 g

CAS Number:  10031-26-2
Price
26,10 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Ferric bromide, Iron tribromide
  • CAS Number:10031-26-2
  • Formula:Br₃Fe
  • MDL Number:MFCD00016082
  • Molecular Weight:295.56 g/mol
  • Melting Pt:200 °C
  • Storage Temperature:Ambient
  • EINECS:233-089-1
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications