0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Iron (III) bromide
Iron(III) bromide, anhydrous
Iron(III) bromide, anhydrous
Bottle, 100 g
APOSIN2056-100G
:
Apollo Scientific
Iron(III) bromide, anhydrous
APOSIN2056-100G
:
Apollo Scientific
:
10031-26-2
Iron(III) bromide, anhydrous
Bottle, 100 g
:
10031-26-2
139,00 €
1 * 100 g
-
+
:
Ferric bromide, Iron tribromide
:
10031-26-2
:
Br₃Fe
:
MFCD00016082
:
295.56 g/mol
:
200 °C
:
Ambient
:
233-089-1
Pack type:
Bottle
Pk:
100 g