Order Entry
Export
Orders LinkContactUsLinkComponent
Zinc molybdate

Zinc molybdate

Bottle, 25 g
Catalog # APOSIN38974-25G
Irritant
Supplier:   Apollo Scientific
Zinc molybdate
Catalog # APOSIN38974-25G
Supplier:  Apollo Scientific
CAS Number:  13767-32-3
Zinc molybdate

Bottle, 25 g

CAS Number:  13767-32-3
Price
387,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Zinc molybdenum oxide
  • CAS Number:13767-32-3
  • Formula:MoO₄Zn
  • MDL Number:MFCD00049622
  • Molecular Weight:225.326 g/mol
  • Melting Pt:>700 °C
  • Melting Pt:>700 °C
  • Density:4.3 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:237-377-8
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications