Order Entry
Export
Orders LinkContactUsLinkComponent
1,9-Pyrazoloanthrone ≥99%

1,9-Pyrazoloanthrone ≥99%

Bottle, 1 g
Catalog # APOSBIS1100-1G
Irritant
Supplier:   Apollo Scientific
1,9-Pyrazoloanthrone ≥99%
Catalog # APOSBIS1100-1G
Supplier:  Apollo Scientific
CAS Number:  129-56-6
1,9-Pyrazoloanthrone ≥99%

Bottle, 1 g

CAS Number:  129-56-6
Price
442,00 €
Log in for availability
1 * 1 g
Product Details & Documents

Novel, potent and selective JNK-1,-2, and -3 inhibitor (Ki = 0.19 uM). SP600125 is an ATP-competitive reversible inhibitor with >20-fold selectivity vs. a range of kinases and enzymes tested. Proc. Natl. Acad. Sci. USA 98: 13681-13686 (2001).

Synonyms:  JNK Inhibitor II, Anthra[1,9-cd]pyrazol-6(2H)-one, Dibenzo[cd,g]indazol-6(2H)-one, SP600125, SP 600125
  • CAS Number:129-56-6
  • Formula:C₁₄H₈N₂O
  • MDL Number:MFCD00022289
  • Molecular Weight:220.231 g/mol
  • Storage Temperature:Freezer
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications