0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Proteins and Peptides
Sheep Recombinant Prl-A
Sheep Recombinant Prl-A
1 mg
APOSBITP1255-1MG
:
Apollo Scientific
Sheep Recombinant Prl-A
APOSBITP1255-1MG
:
Apollo Scientific
:
Sheep Recombinant Prl-A
1 mg
:
1 910,00 €
1 * 1 mg
-
+
Pk:
1 mg
Cat. No.:
APOSBITP1255-1MG
Protein/peptide type:
Recombinant
Protein/peptide name:
Prl-A
Protein synonyms:
Prolactin Antagonist
Species:
Sheep