Order Entry
Export
Orders LinkContactUsLinkComponent
 

Human Recombinant Prl

1 mg
Catalog # APOSBITP1245-1MG
Supplier:   Apollo Scientific
Human Recombinant Prl
Catalog # APOSBITP1245-1MG
Supplier:  Apollo Scientific
CAS Number:  
Human Recombinant Prl

1 mg

CAS Number:  
Price
1 900,00 €
Log in for availability
1 * 1 mg
Product Details & Documents
Specifications

Specifications

  • Pk:
    1 mg
  • Cat. No.:
    APOSBITP1245-1MG
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    Prl
  • Protein synonyms:
    Prolactin
  • Species:
    Human

Specifications