3-Hydroxypropionic acid 30% in aqueous solution
Bottle, 10 g
Catalog # APOSOR59874-10G
Supplier: Apollo Scientific
3-Hydroxypropionic acid 30% in aqueous solution
Catalog # APOSOR59874-10G
Supplier: Apollo Scientific
CAS Number: 503-66-2
Jump to:
Product Details & Documents
Synonyms: 3-Hydroxypropionic Acid, 3-Hydroxypropionic Acid (contains varying amounts of 3,3-Oxydipropionic Acid), Hydracrylic Acid
- CAS Number:503-66-2
- Formula:C₃H₆O₃
- MDL Number:MFCD00058998
- Molecular Weight:90.08 g/mol
- Density:1.08 g/cm³ (25 °C)
- Storage Temperature:Refrigerator
- EINECS:207-974-8
Specifications
Specifications
- Pack type:Bottle
- Pk:10 g



