Uridine 99%
Bottle, 250 g
Catalog # APOSBIU0400-250G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 1-β-D-Ribofuranosyluracil, Uracil-1-β-D-ribofuranoside
- CAS Number:58-96-8
- Formula:C₉H₁₂N₂O₆
- MDL Number:MFCD00006526
- Molecular Weight:244.20 g/mol
- Melting Pt:163…169 °C
- Storage Temperature:Refrigerator
- EINECS:200-407-5
Specifications
Specifications
- Pack type:Bottle
- Pk:250 g



