Order Entry
Export
Orders LinkContactUsLinkComponent
1-Docosene

1-Docosene

Bottle, 100 g
Catalog # APOSOR460033-100G
Supplier:   Apollo Scientific
1-Docosene
Catalog # APOSOR460033-100G
Supplier:  Apollo Scientific
CAS Number:  1599-67-3
1-Docosene

Bottle, 100 g

CAS Number:  1599-67-3
Price
575,00 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:1599-67-3
  • Formula:C₂₂H₄₄
  • MDL Number:MFCD00027085
  • Molecular Weight:308.59 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications