Order Entry
Export
Orders LinkContactUsLinkComponent
2,3-Difluorophenol 98%

2,3-Difluorophenol 98%

Bottle, 5 g
Catalog # APOSPC2865-5G
Flammable
Irritant
Supplier:   Apollo Scientific
2,3-Difluorophenol 98%
Catalog # APOSPC2865-5G
Supplier:  Apollo Scientific
CAS Number:  6418-38-8
2,3-Difluorophenol 98%

Bottle, 5 g

CAS Number:  6418-38-8
Price
21,80 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:6418-38-8
  • Formula:C₆H₄F₂O
  • MDL Number:MFCD00010262
  • Molecular Weight:130.09 g/mol
  • Boiling Pt:53…55 °C/25mmHg
  • Melting Pt:34…38 °C
  • Flash Pt:56 °C
  • Storage Temperature:Ambient
  • UN:1325
  • ADR:4.1,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications