Thymidine (2'-deoxythymidine)
Bottle, 25 g
APOSBIT2044-25G
: 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione, 1-(2-Deoxy-β-D-ribofuranosyl)thymine, 1-(2-Deoxy-β-D-ribofuranosyl)-5-methyluracil, dT, Thymine deoxyriboside, 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione, Pyrimidine deoxynucleoside, Thymidine kinase, Thymidine-2-deoxythymidine
- :50-89-5
- :C₁₀H₁₄N₂O₅
- :MFCD00006537
- :242.23 g/mol
- :186…188 °C
- :Ambient
- :200-070-4
- Pack type:Bottle
- Pk:25 g