Tobramycin
Bottle, 500 mg
Catalog # APOSBIT0802-500MG
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Tobramycin is an aminoglycoside antibiotic and hasa bactericidal action against many gram-negative bacteria. Aminoglycosides are taken into sensitivebacterial cells by an active transport process.
Synonyms: (2S,3R,4S,5S,6R)-4-Amino-2-(((1S,2S,3R,4S,6R)-4,6-diamino-3-(((2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxytetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,5-diol
- CAS Number:32986-56-4
- Formula:C₁₈H₃₇N₅O₉
- MDL Number:MFCD00077885
- Molecular Weight:467.51 g/mol
- Storage Temperature:Refrigerator
- EINECS:251-322-5
Specifications
Specifications
- Pack type:Bottle
- Pk:500 mg



