0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
L(-)-Tryptophan
L(-)-Tryptophan ≥99.5%
L(-)-Tryptophan ≥99.5%
L(-)-Tryptophan ≥99.5%
Bottle, 100 g
APOSBIT0720-100G
:
Apollo Scientific
L(-)-Tryptophan ≥99.5%
APOSBIT0720-100G
:
Apollo Scientific
:
73-22-3
Expand image
L(-)-Tryptophan ≥99.5%
Bottle, 100 g
Add to cart
:
73-22-3
21,80 €
Unit of Measure
1 * 100 g
-
+
Add to cart
USP, pyrogen free
:
L-Tryptophan, Tryptophan, L-α-Amino-3-indolepropionic acid, (S)-2-Amino-3-(3-indolyl)propionic acid, H-Trp-OH
:
73-22-3
:
C₁₁H₁₂N₂O₂
:
MFCD00064340
:
204.23 g/mol
:
280…285 °C (dec.)
:
Ambient
:
200-795-6
Documents
Pack type:
Bottle
Pk:
100 g