3,5-Dihydroxybenzaldehyde
Bottle, 1 g
Catalog # APOSOR41006-1G
Supplier: Apollo Scientific
3,5-Dihydroxybenzaldehyde
Catalog # APOSOR41006-1G
Supplier: Apollo Scientific
CAS Number: 26153-38-8
Jump to:
Product Details & Documents
- CAS Number:26153-38-8
- Formula:C₇H₆O₃
- MDL Number:MFCD00016611
- Molecular Weight:138.12 g/mol
- Melting Pt:153…161 °C
- Storage Temperature:Ambient
- EINECS:247-479-4
Specifications
Specifications
- Pack type:Bottle
- Pk:1 g



