Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,3-Diiodopropane

Bottle, 25 g
Catalog # APOSOR4510-25G
Irritant
Supplier:   Apollo Scientific
1,3-Diiodopropane
Catalog # APOSOR4510-25G
Supplier:  Apollo Scientific
CAS Number:  627-31-6
1,3-Diiodopropane

Bottle, 25 g

CAS Number:  627-31-6
Price
24,60 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Trimethylene diiodide
  • CAS Number:627-31-6
  • Formula:C₃H₆I₂
  • MDL Number:MFCD00001097
  • Molecular Weight:295.89 g/mol
  • Boiling Pt:111…113 °C/23mmHg
  • Melting Pt:–20 °C
  • Density:2.576 g/cm³ (25 °C)
  • Flash Pt:>110 °C
  • Storage Temperature:Refrigerator
  • EINECS:210-993-4
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications