Order Entry
Export
Orders LinkContactUsLinkComponent
2,5-Dichloropyridine

2,5-Dichloropyridine

Bottle, 25 g
Catalog # APOSOR4274-25G
Irritant
Supplier:   Apollo Scientific
2,5-Dichloropyridine
Catalog # APOSOR4274-25G
Supplier:  Apollo Scientific
CAS Number:  16110-09-1
2,5-Dichloropyridine

Bottle, 25 g

CAS Number:  16110-09-1
Price
24,60 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:16110-09-1
  • Formula:C₅H₃Cl₂N
  • MDL Number:MFCD00006239
  • Molecular Weight:147.99 g/mol
  • Boiling Pt:190…191 °C
  • Melting Pt:59…62 °C
  • Flash Pt:112 °C
  • Storage Temperature:Ambient
  • EINECS:240-278-2
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications