Coronene
Bottle, 1 g
Catalog # APOSOR40595-1G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: Hexabenzobenzene
- CAS Number:191-07-1
- Formula:C₂₄H₁₂
- MDL Number:MFCD00004134
- Molecular Weight:300.36 g/mol
- Boiling Pt:525 °C
- Melting Pt:428 °C
- Storage Temperature:Ambient
- EINECS:205-881-7
Specifications
Specifications
- Pack type:Bottle
- Pk:1 g



