0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
1,6-Diiodoperfluorohexane
1,6-Diiodoperfluorohexane 98%
1,6-Diiodoperfluorohexane 98%
Bottle, 5 g
APOSPC2967-5G
:
Apollo Scientific
1,6-Diiodoperfluorohexane 98%
APOSPC2967-5G
:
Apollo Scientific
:
375-80-4
1,6-Diiodoperfluorohexane 98%
Bottle, 5 g
:
375-80-4
181,00 €
1 * 5 g
-
+
:
Perfluoro-1,6-diiodohexane
:
375-80-4
:
C₆F₁₂I₂
:
MFCD00042264
:
553.86 g/mol
:
114…116 °C/100mmHg
:
25 °C
:
2.357 g/cm³ (25 °C)
:
Refrigerator
:
206-794-7
Pack type:
Bottle
Pk:
5 g