Order Entry
Export
Orders LinkContactUsLinkComponent
1,6-Diiodoperfluorohexane 98%

1,6-Diiodoperfluorohexane 98%

Bottle, 25 g
Catalog # APOSPC2967-25G
Irritant
Supplier:   Apollo Scientific
1,6-Diiodoperfluorohexane 98%
Catalog # APOSPC2967-25G
Supplier:  Apollo Scientific
CAS Number:  375-80-4
1,6-Diiodoperfluorohexane 98%

Bottle, 25 g

CAS Number:  375-80-4
Price
724,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Perfluoro-1,6-diiodohexane
  • CAS Number:375-80-4
  • Formula:C₆F₁₂I₂
  • MDL Number:MFCD00042264
  • Molecular Weight:553.86 g/mol
  • Boiling Pt:114…116 °C/100mmHg
  • Melting Pt:25 °C
  • Density:2.357 g/cm³ (25 °C)
  • Storage Temperature:Refrigerator
  • EINECS:206-794-7
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications