Linoleic acid
Bottle, 25 g
Catalog # APOSBIL9075-25G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: (9Z,12Z)-Octadeca-9,12-dienoic acid, cis,cis-9,12-Octadecadienoic acid, 9Z,12Z-Octadecadienoic acid
- CAS Number:60-33-3
- Formula:C₁₈H₃₂O₂
- MDL Number:MFCD00064241
- Molecular Weight:280.45 g/mol
- Boiling Pt:230 °C/15mmHg
- Melting Pt:–12 °C
- Density:0.902 g/cm³ (25 °C)
- Flash Pt:>110 °C
- Storage Temperature:Freezer
- EINECS:200-470-9
Specifications
Specifications
- Pack type:Bottle
- Pk:25 g



