Order Entry
Export
Orders LinkContactUsLinkComponent
Linoleic acid

Linoleic acid

Bottle, 10 g
Catalog # APOSBIL9075-10G
Irritant
Supplier:   Apollo Scientific
Linoleic acid
Catalog # APOSBIL9075-10G
Supplier:  Apollo Scientific
CAS Number:  60-33-3
Linoleic acid

Bottle, 10 g

CAS Number:  60-33-3
Price
49,20 €
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  (9Z,12Z)-Octadeca-9,12-dienoic acid, cis,cis-9,12-Octadecadienoic acid, 9Z,12Z-Octadecadienoic acid
  • CAS Number:60-33-3
  • Formula:C₁₈H₃₂O₂
  • MDL Number:MFCD00064241
  • Molecular Weight:280.45 g/mol
  • Boiling Pt:230 °C/15mmHg
  • Melting Pt:–12 °C
  • Density:0.902 g/cm³ (25 °C)
  • Flash Pt:>110 °C
  • Storage Temperature:Freezer
  • EINECS:200-470-9
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g

Specifications