Order Entry
Export
Orders LinkContactUsLinkComponent
2,3-Dibromopyridine

2,3-Dibromopyridine

Bottle, 100 g
Catalog # APOSOR4486-100G
Irritant
Supplier:   Apollo Scientific
2,3-Dibromopyridine
Catalog # APOSOR4486-100G
Supplier:  Apollo Scientific
CAS Number:  13534-89-9
2,3-Dibromopyridine

Bottle, 100 g

CAS Number:  13534-89-9
Price
218,00 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:13534-89-9
  • Formula:C₅H₃Br₂N
  • MDL Number:MFCD00234014
  • Molecular Weight:236.89 g/mol
  • Boiling Pt:249…250 °C
  • Melting Pt:56…60 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications