Order Entry
Export
Orders LinkContactUsLinkComponent
Nicotinamide 99%

Nicotinamide 99%

Bottle, 250 g
Catalog # APOSBIN0610-250G
Irritant
Supplier:   Apollo Scientific
Nicotinamide 99%
Catalog # APOSBIN0610-250G
Supplier:  Apollo Scientific
CAS Number:  98-92-0
Nicotinamide 99%

Bottle, 250 g

CAS Number:  98-92-0
Price
36,20 €
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  3-(Aminocarbonyl)pyridine, Niacinamide, pyridine-3-carboxamide, Vitamin B3
  • CAS Number:98-92-0
  • Formula:C₆H₆N₂O
  • MDL Number:MFCD00006395
  • Molecular Weight:122.13 g/mol
  • Melting Pt:128…132 °C
  • Density:1.4 g/cm³ (25 °C)
  • Flash Pt:150 °C
  • Storage Temperature:Ambient
  • EINECS:202-713-4
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 g

Specifications