Order Entry
Export
Orders LinkContactUsLinkComponent
 

Basic blue 12

Bottle, 25 g
Catalog # APOSBIN0464-25G
Irritant
Supplier:   Apollo Scientific
Basic blue 12
Catalog # APOSBIN0464-25G
Supplier:  Apollo Scientific
CAS Number:  2381-85-3
Basic blue 12

Bottle, 25 g

CAS Number:  2381-85-3
Price
72,40 €
Log in for availability
1 * 25 g
Product Details & Documents

Fluorecence reference standard.

Synonyms:  Nile Blue A chloride, 5-Amino-9-(diethylamino)benzo[a]phenoxazin-7-ium chloride
  • CAS Number:2381-85-3
  • Formula:C₂₀H₂₀ClN₃O
  • MDL Number:MFCD00011936
  • Molecular Weight:353.85 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications