Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Difluorobenzene 99%

1,3-Difluorobenzene 99%

Bottle, 2,5 kg
Catalog # APOSPC2630-2.5KG
Flammable
Irritant
Supplier:   Apollo Scientific
1,3-Difluorobenzene 99%
Catalog # APOSPC2630-2.5KG
Supplier:  Apollo Scientific
CAS Number:  372-18-9
1,3-Difluorobenzene 99%

Bottle, 2,5 kg

CAS Number:  372-18-9
Price
138,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents
  • CAS Number:372-18-9
  • Formula:C₆H₄F₂
  • MDL Number:MFCD00000327
  • Molecular Weight:114.09 g/mol
  • Boiling Pt:82 °C
  • Melting Pt:–59 °C
  • Density:1.163 g/cm³ (25 °C)
  • Flash Pt:2 °C
  • Storage Temperature:Ambient
  • EINECS:206-746-5
  • UN:1993
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    2,5 kg

Specifications