Nalidixic acid
Bottle, 25 g
Catalog # APOSBIN0134-25G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Active against gram-negative bacteria.
- CAS Number:389-08-2
- Formula:C₁₂H₁₂N₂O₃
- MDL Number:MFCD00006884
- Molecular Weight:232.24 g/mol
- Melting Pt:225…230 °C
- Storage Temperature:Ambient
- EINECS:206-864-7
Specifications
Specifications
- Pack type:Bottle
- Pk:25 g



