Order Entry
Export
Orders LinkContactUsLinkComponent
Uric acid

Uric acid

Bottle, 500 g
Catalog # APOSBIK9021-500G
Irritant
Supplier:   Apollo Scientific
Uric acid
Catalog # APOSBIK9021-500G
Supplier:  Apollo Scientific
CAS Number:  69-93-2
Uric acid

Bottle, 500 g

CAS Number:  69-93-2
Price
420,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  1H-Purine-2,6,8(3H,7H,9H)-trione
  • CAS Number:69-93-2
  • Formula:C₅H₄N₄O₃
  • MDL Number:MFCD00005712
  • Molecular Weight:168.11 g/mol
  • Melting Pt:>300 °C
  • Melting Pt:>300 °C
  • Storage Temperature:Ambient
  • EINECS:200-720-7
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g

Specifications