Order Entry
Export
Orders LinkContactUsLinkComponent
Benzophenone imine 97%

Benzophenone imine 97%

Bottle, 500 g
Catalog # APOSOR42185-500G
Irritant
Supplier:   Apollo Scientific
Benzophenone imine 97%
Catalog # APOSOR42185-500G
Supplier:  Apollo Scientific
CAS Number:  1013-88-3
Benzophenone imine 97%

Bottle, 500 g

CAS Number:  1013-88-3
Price
197,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Benzophenoneimine
  • CAS Number:1013-88-3
  • Formula:C₁₃H₁₁N
  • MDL Number:MFCD00001760
  • Molecular Weight:181.24 g/mol
  • Boiling Pt:151…153 °C/10mmHg
  • Density:1.08 g/cm³ (25 °C)
  • Flash Pt:109 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g

Specifications