Diacetoxyscirpenol Standard Solution
Diacetoxyscirpenol, 100 µg/ml in acetonitrile, Bottle, 1 mL
Catalog # APOSBIMS125-1ML
Supplier: Apollo Scientific
Diacetoxyscirpenol Standard Solution
Catalog # APOSBIMS125-1ML
Supplier: Apollo Scientific
CAS Number:
Jump to:
Product Details & Documents
Analytical reference standard solution in acetonitrile.
Caution: Expires in 12 months.
Specifications
Specifications
- Concentration:100 µg/ml in acetonitrile
- Pack type:Bottle
- Pk:1 mL
- Description:Diacetoxyscirpenol
- Cat. No.:APOSBIMS125-1ML
- Formula:C₁₉H₂₆O₇
- Description:Diacetoxyscirpenol
- Storage temperature:Freezer
- Flash point:2 °C
- CAS No.:2270-40-8



