Zearalenone standard solution
Zearalenone, 100 µg/ml in acetonitrile, Bottle, 1 mL
Catalog # APOSBIMS119-1ML
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Analytical reference standard solution in acetonitrile.
Caution: Expires in 12 months.
Specifications
Specifications
- Concentration:100 µg/ml in acetonitrile
- Pack type:Bottle
- Pk:1 mL
- Description:Zearalenone
- Cat. No.:APOSBIMS119-1ML
- Formula:C₁₈H₂₂O₅
- Description:Zearalenone
- Boiling point:81…82 °C
- Storage temperature:Freezer
- Flash point:2 °C
- CAS No.:17924-92-4



