T2 Triol standard solution
T2 Triol, 100 µg/ml in acetonitrile, Bottle, 5 mL
APOSBIMS113-5ML
Analytical reference standard solution in acetonitrile.
: Expires in 12 months.
- Concentration:100 µg/ml in acetonitrile
- Pack type:Bottle
- Pk:5 mL
- Description:T2 Triol
- Cat. No.:APOSBIMS113-5ML
- Formula:C₂₀H₃₀O₇
- Description:T2 Triol
- Storage temperature:Freezer
- Flash point:2 °C
- CAS No.:34114-98-2



