T2 Triol standard solution
T2 Triol, 100 µg/ml in acetonitrile, Bottle, 1 mL
Catalog # APOSBIMS113-1ML
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Analytical reference standard solution in acetonitrile.
Caution: Expires in 12 months.
Specifications
Specifications
- Concentration:100 µg/ml in acetonitrile
- Pack type:Bottle
- Pk:1 mL
- Description:T2 Triol
- Cat. No.:APOSBIMS113-1ML
- Formula:C₂₀H₃₀O₇
- Description:T2 Triol
- Storage temperature:Freezer
- Flash point:2 °C
- CAS No.:34114-98-2
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



